2,5-Diethynylpyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D4664-250MG D4664-1G
TCIAD4664-250MGEA 68.2 EUR
TCIAD4664-250MG TCIAD4664-1G
2,5-Diethynylpyridine ≥98.0% (by GC)
2,5-Diethynylpyridine
Formule: C₉H₅N
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD11109672
CAS nummer: 137000-75-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR