2,7-Diamino-9,9-di-n-octylfluorene ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 9,9-Dioctyl-9H-fluorene-2,7-diamine

D4719-200MG D4719-1G
TCIAD4719-200MGEA 86.9 EUR
TCIAD4719-200MG TCIAD4719-1G
2,7-Diamino-9,9-di-n-octylfluorene ≥98.0% (by HPLC, titration analysis)
2,7-Diamino-9,9-di-n-octylfluorene
Formule: C₂₉H₄₄N₂
MW: 420.69 g/mol
MDL Nummer: MFCD27923070
CAS nummer: 851042-10-9

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR