L-Glutamic acid dibenzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D4839-5G D4839-25G
TCIAD4839-5GEA 37.4 EUR
TCIAD4839-5G TCIAD4839-25G
L-Glutamic acid dibenzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
L-Glutamic acid dibenzyl ester hydrochloride
Formule: C₁₉H₂₁NO₄·HCl
MDL Nummer: MFCD00035103
CAS nummer: 4561-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3493308244eab3b517c993028a37fe2af62c553&vid=d3493308244eab3b517c993028a37fe2af62c553&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 102.0 to 106.0 deg-C
Specific rotation [a]20/D +8.5 to +10.0 deg (C=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR