9,9-Dimethylfluorene-2-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D5018-1G D5018-5G
TCIAD5018-1GEA 29.7 EUR
TCIAD5018-1G TCIAD5018-5G
9,9-Dimethylfluorene-2-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
9,9-Dimethylfluorene-2-boronic acid pinacol ester
Formule: C₂₁H₂₅BO₂
MDL Nummer: MFCD08704229
CAS nummer: 569343-09-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR