2,4-Dichloro-3-formylpyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D5076-1G D5076-5G
TCIAD5076-1GEA 95.7 EUR
TCIAD5076-1G TCIAD5076-5G
2,4-Dichloro-3-formylpyridine ≥98.0% (by GC)
2,4-Dichloro-3-formylpyridine
Formule: C₆H₃Cl₂NO
MDL Nummer: MFCD07437909
CAS nummer: 134031-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=def09b8ee40cc9014044d0352994b4bf3ab4edec&vid=def09b8ee40cc9014044d0352994b4bf3ab4edec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR