2,6-Difluoro-3-nitrobenzonitrile ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D5185-1G D5185-5G
TCIAD5185-1GEA 17.6 EUR
TCIAD5185-1G TCIAD5185-5G
2,6-Difluoro-3-nitrobenzonitrile ≥97.0% (by GC)
2,6-Difluoro-3-nitrobenzonitrile
Formule: C₇H₂F₂N₂O₂
MDL Nummer: MFCD00042254
CAS nummer: 143879-77-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR