4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D5200-1G D5200-5G
TCIAD5200-1GEA 56.1 EUR
TCIAD5200-1G TCIAD5200-5G
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane ≥95.0% (by GC)
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
Formule: C₇H₁₂O₃
Kookpunt: 86°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD16621162
CAS nummer: 57280-22-5
EINECS: 421-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105369269d418cc7aedf0db7edb29ec4ccb70760&vid=105369269d418cc7aedf0db7edb29ec4ccb70760&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR