1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D5316-1G D5316-5G
TCIAD5316-1GEA 121 EUR
TCIAD5316-1G TCIAD5316-5G
1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)
1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene
Formule: C₁₆H₂₀Br₂Si₂
MDL Nummer: MFCD11521283
CAS nummer: 478190-79-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR