4,6-Di(tert-butyl)benzene-1,3-diol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4,6-Di-tert-butylresorcinol, 4,6-Di-tert-butylbenzene-1,3-diol

D5459-25G D5459-5G
TCIAD5459-25GEA 220 EUR
TCIAD5459-25G TCIAD5459-5G
4,6-Di(tert-butyl)benzene-1,3-diol ≥98.0% (by GC)
4,6-Di(tert-butyl)benzene-1,3-diol
Formule: C₁₄H₂₂O₂
MW: 222.33 g/mol
MDL Nummer: MFCD00044513
CAS nummer: 5374-06-1
EINECS: 226-366-3

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR