4,4'-Dimethylbenzhydrol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Bis(4-methylphenyl)methanol, 4,4'-Dimethylbenzhydryl alcohol, Di-p-tolylmethanol, 4,4'-Dimethyl-α-phenylbenzyl alcohol, Bis(4-methylphenyl)carbinol, 4,4'-Dimethylphenyl phenyl carbinol, 4,4'-Dimethyldiphenylmethanol

D5460-1G D5460-5G
TCIAD5460-1GEA 71.5 EUR
TCIAD5460-1G TCIAD5460-5G
4,4'-Dimethylbenzhydrol ≥98.0% (by GC)
4,4'-Dimethylbenzhydrol
Formule: C₁₅H₁₆O
MW: 212.29 g/mol
MDL Nummer: MFCD00017216
CAS nummer: 885-77-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5768452a85e9962920656368ba890f6bc65d1630&vid=5768452a85e9962920656368ba890f6bc65d1630&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR