Mesityleneruthenium(II) chloride Dimer ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D5524-250MG D5524-1G
TCIAD5524-250MGEA 88 EUR
TCIAD5524-250MG TCIAD5524-1G
Mesityleneruthenium(II) chloride Dimer ≥97.0% (by titrimetric analysis)
Mesityleneruthenium(II) chloride Dimer
Formule: C₁₈H₂₄Cl₄Ru₂
MDL Nummer: MFCD11656079
CAS nummer: 52462-31-4

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Argentmetric Titration) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR