N,N-Dimethyl-1,3-propanediamine dihydroiodide ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D5619-1G D5619-5G
TCIAD5619-1GEA 31.9 EUR
TCIAD5619-1G TCIAD5619-5G
N,N-Dimethyl-1,3-propanediamine dihydroiodide ≥97.0% (by titrimetric analysis)
N,N-Dimethyl-1,3-propanediamine dihydroiodide
Formule: C₅H₁₄N₂·₂HI
CAS nummer: 2561497-43-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR