2,5-Dibromo-3-fluorothiophene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D5729-1G D5729-5G
TCIAD5729-1GEA 156 EUR
TCIAD5729-1G TCIAD5729-5G
2,5-Dibromo-3-fluorothiophene ≥98.0% (by GC)
2,5-Dibromo-3-fluorothiophene
Formule: C₄HBr₂FS
Kookpunt: 78°C
Bewaar temperatuur: Vriezer
CAS nummer: 32431-85-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR