Ethylmagnesium bromide 13% (w/w) ∼1 mol/L in THF

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

E0497-250G
TCIAE0497-250GEA 92.4 EUR
TCIAE0497-250G
Ethylmagnesium bromide 13% (w/w) ∼1 mol/L in THF
Ethylmagnesium bromide
Formule: C₂H₅BrMg
MDL Nummer: MFCD00000043
CAS nummer: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3415519824b838fe0cecea9bd5a406862d8b1fae&vid=3f8906750c08178ded8e86a3a1c491b83831056f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Concentration (sec Butanol method) 13.0 to 14.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR