1-Ethyl-4-piperidone ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

E0498-25ML E0498-100ML
TCIAE0498-25MLEA 97.9 EUR
TCIAE0498-25ML TCIAE0498-100ML
1-Ethyl-4-piperidone ≥98.0% (by GC, titration analysis)
1-Ethyl-4-piperidone
Formule: C₇H₁₃NO
Kookpunt: 80°C
Vlampunt: 68°C
MDL Nummer: MFCD00013356
CAS nummer: 3612-18-8
EINECS: 222-781-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e12004953686f418b0326bc52bb26cf051a7283&vid=3e12004953686f418b0326bc52bb26cf051a7283&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR