1-Ethyl-3-methylimidazolium ethylsulphate ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

E0650-25G E0650-5G
TCIAE0650-25GEA 79.2 EUR
TCIAE0650-25G TCIAE0650-5G
1-Ethyl-3-methylimidazolium ethylsulphate ≥98.0% (by total nitrogen basis)
1-Ethyl-3-methylimidazolium ethylsulphate
Formule: C₈H₁₆N₂O₄S
Vlampunt: 162°C
MDL Nummer: MFCD06798189
CAS nummer: 342573-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14adbf116dabf419509d73a392f21ec9883ef10f&vid=14adbf116dabf419509d73a392f21ec9883ef10f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (with Total Nitrogen) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR