Ethyl-3-hydroxyhexanoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Ethyl 3-hydroxyhexanoate

E0788-100G E0788-25G
TCIAE0788-100GEA 196 EUR
TCIAE0788-100G TCIAE0788-25G
Ethyl-3-hydroxyhexanoate ≥98.0% (by GC)
Ethyl-3-hydroxyhexanoate
Formule: C₈H₁₆O₃
MW: 160.21 g/mol
Kookpunt: 92°C
Vlampunt: 94°C
MDL Nummer: MFCD00036604
CAS nummer: 2305-25-1
EINECS: 218-973-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9bcac6dba02f5985013211de18d491da362e9ad&vid=a9bcac6dba02f5985013211de18d491da362e9ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR