Imidazole-4-carboxylic acid ethyl ester ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

E1042-1G E1042-5G
TCIAE1042-1GEA 19.8 EUR
TCIAE1042-1G TCIAE1042-5G
Imidazole-4-carboxylic acid ethyl ester ≥98.0% (by GC, titration analysis)
Imidazole-4-carboxylic acid ethyl ester
Formule: C₆H₈N₂O₂
MDL Nummer: MFCD07363812
CAS nummer: 23785-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34050a89b6f95fca68ed48c34e6c2aee875407e0&vid=dc10d06d99aa351c0293ebd71f1cc74df09ccd10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 158.0 to 163.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR