Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

E1191-25G E1191-5G
TCIAE1191-25GEA 365 EUR
TCIAE1191-25G TCIAE1191-5G
Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate ≥98.0% (by GC)
Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate
Formule: C₈H₁₀F₂N₂O₂
Kookpunt: 120°C
MDL Nummer: MFCD12022341
CAS nummer: 141573-95-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR