Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

E1283-1G E1283-5G
TCIAE1283-1GEA 49.5 EUR
TCIAE1283-1G TCIAE1283-5G
Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Ethyl(dimethyl)(2-phenylethyl)ammonium bis(trifluoromethanesulphonyl)imide
Formule: C₁₄H₂₀F₆N₂O₄S₂
CAS nummer: 1804970-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41f1fc856dd63055a4c41b4e2d2a52dd940abcff&vid=41f1fc856dd63055a4c41b4e2d2a52dd940abcff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (HPLC) min. 96.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR