Etretinate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

E1293-50MG E1293-250MG
TCIAE1293-50MGEA 75.9 EUR
TCIAE1293-50MG TCIAE1293-250MG
Etretinate ≥98.0% (by HPLC)
Etretinate
Formule: C₂₃H₃₀O₃
MDL Nummer: MFCD00866624
CAS nummer: 54350-48-0
EINECS: 259-119-3
UN: 3077
ADR: 9,III
Merck Index: 14,03892
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b60a80c37dc860ed8b521decb878913363f74245&vid=b60a80c37dc860ed8b521decb878913363f74245&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR