Ethyl trimethylacetopyruvate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

E1326-1G E1326-5G
TCIAE1326-1GEA 57.2 EUR
TCIAE1326-1G TCIAE1326-5G
Ethyl trimethylacetopyruvate ≥98.0% (by GC)
Ethyl trimethylacetopyruvate
Formule: C₁₀H₁₆O₄
Kookpunt: 124°C
MDL Nummer: MFCD00052319
CAS nummer: 13395-36-3
EINECS: 236-478-4

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR