4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

E1393-1G
TCIAE1393-1GEA 118 EUR
TCIAE1393-1G
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester ≥98.0% (by titrimetric analysis)
4-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester
Formule: C₁₆H₂₃BO₄
MDL Nummer: MFCD08704684
CAS nummer: 859169-20-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR