FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

F0026-1G F0026-100MG
TCIAF0026-1GEA 341 EUR
TCIAF0026-1G TCIAF0026-100MG
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥97.0% (by HPLC, titration analysis)
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Formule: C₂₁H₁₁NO₅S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005063
CAS nummer: 3326-32-7
EINECS: 222-042-0

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR