Fumaarzuur ≥99.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

F0067-25G F0067-500G
TCIAF0067-25GEA 17.6 EUR
TCIAF0067-25G TCIAF0067-500G
Fumaarzuur ≥99.0% (by GC, titration analysis)
Fumaarzuur
Formule: C₄H₄O₄
Kookpunt: 356 °C (1013 hPa)
Smeltpunt: 287°C
Dichtheid: 1,625 g/cm³ (20 °C)
Vlampunt: 273 °C
MDL Nummer: MFCD00002700
CAS nummer: 110-17-8
EINECS: 203-743-0
Merck Index: 14,04287
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e21f7e9afb6b200e1efd8a975187627f1ea2bf00&vid=e21f7e9afb6b200e1efd8a975187627f1ea2bf00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR