Ethyl-2-furoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

F0098-25G
TCIAF0098-25GEA 17.6 EUR
TCIAF0098-25G
Ethyl-2-furoate ≥98.0% (by GC)
Ethyl-2-furoate
Formule: C₇H₈O₃
Kookpunt: 196°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003237
CAS nummer: 614-99-3
EINECS: 210-404-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2df20d5717161f4ddf490fd4b9e6c394a3e6d443&vid=2df20d5717161f4ddf490fd4b9e6c394a3e6d443&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR