N-Formyl-L-leucine

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

F0131-1G F0131-5G
TCIAF0131-1GEA 34.1 EUR
TCIAF0131-1G TCIAF0131-5G
N-Formyl-L-leucine
N-Formyl-L-leucine
Formule: C₇H₁₃NO₃
MDL Nummer: MFCD00055861
CAS nummer: 6113-61-7
EINECS: 228-080-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87a754ed1c2fc4a756e0072ba0389a165c041951&vid=87a754ed1c2fc4a756e0072ba0389a165c041951&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 141.0 to 144.0 deg-C
Specific rotation [a]20/D -38.0 to -41.0 deg (C=1, H2O)
Solubility in hot Water almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR