α-Bromo-2-fluorotoulene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-Fluorobenzyl bromide, α-Bromo-o-fluorotoulene

F0209-25G F0209-5G
TCIAF0209-25GEA 64.9 EUR
TCIAF0209-25G TCIAF0209-5G
α-Bromo-2-fluorotoulene ≥98.0% (by GC)
α-Bromo-2-fluorotoulene
Formule: C₇H₆BrF
MW: 189.03 g/mol
Kookpunt: 115°C
Vlampunt: 82°C
MDL Nummer: MFCD00000324
CAS nummer: 446-48-0
EINECS: 207-169-1
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8cdf36ed35740b4794027d0559660447bfceec2&vid=a2d437a8c27923d4ff96e3b4efc29c2db65d3699&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR