N-Fmoc-glycine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

F0293-25G F0293-5G
TCIAF0293-25GEA 67.1 EUR
TCIAF0293-25G TCIAF0293-5G
N-Fmoc-glycine ≥98.0% (by HPLC, titration analysis)
N-Fmoc-glycine
Formule: C₁₇H₁₅NO₄
MW: 297.3 g/mol
MDL Nummer: MFCD00037140
CAS nummer: 29022-11-5
EINECS: 249-373-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2086f58118ccfe051d61b8b2003078d9e7959bbf&vid=2086f58118ccfe051d61b8b2003078d9e7959bbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 178.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR