N-Fmoc-L-alanine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

F0305-25G F0305-5G
TCIAF0305-25GEA 80.3 EUR
TCIAF0305-25G TCIAF0305-5G
N-Fmoc-L-alanine ≥98.0% (by HPLC, titration analysis)
N-Fmoc-L-alanine
Formule: C₁₈H₁₇NO₄
MDL Nummer: MFCD00037139
CAS nummer: 35661-39-3
EINECS: 252-660-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -17.0 to -21.0 deg (C=1, DMF)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR