4-Fluoro-3-nitrobenzotrifluoride ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

F0324-25G F0324-250G
TCIAF0324-25GEA 17.6 EUR
TCIAF0324-25G TCIAF0324-250G
4-Fluoro-3-nitrobenzotrifluoride ≥95.0% (by GC)
4-Fluoro-3-nitrobenzotrifluoride
Formule: C₇H₃F₄NO₂
Kookpunt: 208°C
Dichtheid: 1,49 g/cm³ (20 °C)
Vlampunt: 33°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007059
CAS nummer: 367-86-2
EINECS: 206-702-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a96b02c80ffd46186c5320b57289d415c25eeb10&vid=d38a8b7343a38635d9a3e5a6fbdca5234195b453&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR