N-Formylglycine ethyl ester ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

F0421-25G F0421-5G
TCIAF0421-25GEA 158 EUR
TCIAF0421-25G TCIAF0421-5G
N-Formylglycine ethyl ester ≥98.0% (by GC)
N-Formylglycine ethyl ester
Formule: C₅H₉NO₃
MDL Nummer: MFCD00014125
CAS nummer: 3154-51-6
EINECS: 221-596-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR