2-(2-Fluorophenyl)ethanol ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

F0778-25G F0778-5G
TCIAF0778-25GEA 113 EUR
TCIAF0778-25G TCIAF0778-5G
2-(2-Fluorophenyl)ethanol ≥95.0% (by GC)
2-(2-Fluorophenyl)ethanol
Formule: C₈H₉FO
Kookpunt: 137°C
Dichtheid: 1,04 g/cm³ (20 °C)
Vlampunt: 98°C
MDL Nummer: MFCD00002887
CAS nummer: 50919-06-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35ba864cc2c6321bb724a8873e128b34da45bf42&vid=35ba864cc2c6321bb724a8873e128b34da45bf42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR