3-Fluoro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

F1064-1G
TCIAF1064-1GEA 115 EUR
TCIAF1064-1G
3-Fluoro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-5-(trifluoromethyl)phenylboronic acid
Formule: C₇H₅BF₄O₂
Smeltpunt: 168°C
MDL Nummer: MFCD06657735
CAS nummer: 159020-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f66dd60f13474fc714bb2b47e457d44de05d35a0&vid=f66dd60f13474fc714bb2b47e457d44de05d35a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 98.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR