Fmoc-N-Me-Ile-OH ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

F1129-1G F1129-5G
TCIAF1129-1GEA 57.2 EUR
TCIAF1129-1G TCIAF1129-5G
Fmoc-N-Me-Ile-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-N-Me-Ile-OH
Formule: C₂₂H₂₅NO₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00153389
CAS nummer: 138775-22-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 177.0 deg-C
Specific rotation [a]20/D -59.0 to -64.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR