trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis), purified by sublimation

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

F1257-1G F1257-5G
TCIAF1257-1GEA 95.7 EUR
TCIAF1257-1G TCIAF1257-5G
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis), purified by sublimation
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
Formule: C₁₀H₁₀O₄
MDL Nummer: MFCD00004400
CAS nummer: 537-98-4
EINECS: 208-679-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR