Gibberellic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G0029-1G G0029-100MG G0029-5G
TCIAG0029-1GEA 33 EUR
TCIAG0029-1G TCIAG0029-100MG TCIAG0029-5G
Gibberellic acid ≥98.0% (by titrimetric analysis)
Gibberellic acid
Formule: C₁₉H₂₂O₆
Smeltpunt: 234 °C
Dichtheid: 1,386 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00079329
CAS nummer: 77-06-5
EINECS: 201-001-0
Merck Index: 14,04419
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1008e1006a13807990c6e095eff7012027addcd3&vid=1008e1006a13807990c6e095eff7012027addcd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +76.0 to +86.0 deg (C=2, Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR