Tripropionin ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G0090-25G G0090-500G
TCIAG0090-25GEA 17.6 EUR
TCIAG0090-25G TCIAG0090-500G
Tripropionin ≥95.0% (by GC)
Tripropionin
Formule: C₁₂H₂₀O₆
Kookpunt: 176°C
Vlampunt: 162°C
MDL Nummer: MFCD00027005
CAS nummer: 139-45-7
EINECS: 205-365-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR