Acid green 3

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G0176-25G
TCIAG0176-25GEA 66 EUR
TCIAG0176-25G
Acid green 3
Acid green 3
Formule: C₃₇H₃₇N₂O₆S₂
MDL Nummer: MFCD00012122
CAS nummer: 4680-78-8
EINECS: 225-132-8
Merck Index: 14,04575

Order Now

Specification Test Results

Appearance Red to Dark blue to Black powder to crystal
Lambda max. 617.0 to 621.0 nm (0.02mol/L AcONH4 sol.)
Absorbance (E1%1cm) min. 600 (0.02mol/L AcONH4 sol., 617.0 to 621.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR