Guanidiniumchloride ≥99.0% (by titrimetric analysis) for protein research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

G0197-25G G0197-500G G0197-100G
TCIAG0197-25GEA 24.2 EUR
TCIAG0197-25G TCIAG0197-500G TCIAG0197-100G
Guanidiniumchloride ≥99.0% (by titrimetric analysis) for protein research
Guanidiniumchloride
Formule: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Smeltpunt: 185
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013026
CAS nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 14,04562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e4dd9f0a4b1455d358721a1fb2ac60050f650c&vid=2fe27eedee6de0a811023e1e664a52fd3375cc70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 185.0 to 189.0 deg-C
Absorbance max. 0.200 (6mol/L, H2O, 225.0 nm)
Suitability for protein-analysis to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR