Calcium glycolate ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G0200-25G
TCIAG0200-25GEA 56.1 EUR
TCIAG0200-25G
Calcium glycolate ≥97.0% (by titrimetric analysis)
Calcium glycolate
Formule: C₂H₄O₃
MDL Nummer: MFCD00067202
CAS nummer: 996-23-6
EINECS: 213-629-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9011daf58fcfa5dd6595da13589625b6ef05660e&vid=9011daf58fcfa5dd6595da13589625b6ef05660e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR