(S)-(+)-Glycidyl butyrate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (S)-Oxiran-2-ylmethyl butyrate

G0283-25G G0283-5G
TCIAG0283-25GEA 165 EUR
TCIAG0283-25G TCIAG0283-5G
(S)-(+)-Glycidyl butyrate ≥98.0% (by GC)
(S)-(+)-Glycidyl butyrate
Formule: C₇H₁₂O₃
MW: 144.1684 g/mol
Vlampunt: 84°C
MDL Nummer: MFCD00273369
CAS nummer: 65031-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d57c90f2c1908960c77526b9f86d928796603a6&vid=3d57c90f2c1908960c77526b9f86d928796603a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +29.0 to +33.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR