2-Heptenoic acid ≥90.0% (by GC) (contains 3-Heptenoic Acid)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H0426-5ML
TCIAH0426-5MLEA 147 EUR
TCIAH0426-5ML
2-Heptenoic acid ≥90.0% (by GC) (contains 3-Heptenoic Acid)
2-Heptenoic acid
Formule: C₇H₁₂O₂
Kookpunt: 122°C
MDL Nummer: MFCD00014019
CAS nummer: 18999-28-5
EINECS: 242-738-8
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR