2-Hydroxy-3-methyl-2-cyclopentenone ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H0469-25G
TCIAH0469-25GEA 57.2 EUR
TCIAH0469-25G
2-Hydroxy-3-methyl-2-cyclopentenone ≥98.0% (by titrimetric analysis)
2-Hydroxy-3-methyl-2-cyclopentenone
Formule: C₆H₈O₂
MDL Nummer: MFCD00013747
CAS nummer: 80-71-7
EINECS: 201-303-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 104.0 to 108.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR