Isobutyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H0493-25G H0493-500G H0493-100G
TCIAH0493-25GEA 30.8 EUR
TCIAH0493-25G TCIAH0493-500G TCIAH0493-100G
Isobutyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis)
Isobutyl-4-hydroxybenzoate
Formule: C₁₁H₁₄O₃
MDL Nummer: MFCD00020167
CAS nummer: 4247-02-3
EINECS: 224-208-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cd9f47d0da18d8ca43c2b02bbb51d7a36530c28&vid=1cd9f47d0da18d8ca43c2b02bbb51d7a36530c28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 75.0 to 79.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR