5-Hydroxy-L-tryptophan ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H0531-1G H0531-5G
TCIAH0531-1GEA 20.9 EUR
TCIAH0531-1G TCIAH0531-5G
5-Hydroxy-L-tryptophan ≥98.0% (by HPLC, titration analysis)
5-Hydroxy-L-tryptophan
Formule: C₁₁H₁₂N₂O₃
Smeltpunt: 273 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00064341
CAS nummer: 4350-09-8
EINECS: 224-411-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,04847
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f24fc82d09087b5b7a340b6b5bec354c2f9f515a&vid=a91f524b9dbb1f37b1a1c1ca5a7aa75d065808dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -31.0 to -35.0 deg (C=1, H2O)
Solubility in hot Water almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR