trans-2-Heptenal ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H0579-25ML H0579-5ML
TCIAH0579-25MLEA 131 EUR
TCIAH0579-25ML TCIAH0579-5ML
trans-2-Heptenal ≥95.0% (by GC)
trans-2-Heptenal
Formule: C₇H₁₂O
Kookpunt: 91°C
Dichtheid: 0,8575 g/cm³ (15 °C)
Vlampunt: 54°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007010
CAS nummer: 18829-55-5
EINECS: 242-608-0
UN: 1988
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=134f0d7f3709e1cf1dc6c342ea793d630d5c0a78&vid=4863443a9f7e0592ecc66baa58a900c772499698&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR