2-Quinoxalinol ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H0591-25G H0591-500G H0591-100G
TCIAH0591-25GEA 23.1 EUR
TCIAH0591-25G TCIAH0591-500G TCIAH0591-100G
2-Quinoxalinol ≥98.0% (by HPLC, titration analysis)
2-Quinoxalinol
Formule: C₈H₆N₂O
MDL Nummer: MFCD00006722
CAS nummer: 1196-57-2
EINECS: 214-815-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed18a3c973f6aced47db106b951a80b05fb0fa24&vid=ed18a3c973f6aced47db106b951a80b05fb0fa24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR