N-(Hydroxymethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H0648-25G H0648-250G
TCIAH0648-25GEA 17.6 EUR
TCIAH0648-25G TCIAH0648-250G
N-(Hydroxymethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(Hydroxymethyl)phthalimide
Formule: C₉H₇NO₃
MDL Nummer: MFCD00005899
CAS nummer: 118-29-6
EINECS: 204-241-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 143.0 to 148.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR