Hexadecyltrimethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H0734-1G H0734-5G
TCIAH0734-1GEA 95.7 EUR
TCIAH0734-1G TCIAH0734-5G
Hexadecyltrimethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Hexadecyltrimethylammonium tetrafluoroborate
Formule: C₁₉H₄₂BF₄N
MDL Nummer: MFCD00144896
CAS nummer: 73257-08-6
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR